Learn More
Med Vet International Water Resistant Dressing Retention Tape With Liner 3M Multipore Dry White 2 inch X 5-1/2 Yard Pique Nonsterile, 3730-2, 48/Case
Catalog No. 502831437
Encompass_Preferred
Reliable securement in damp areas; suitable for wet or diaphoretic skin Bi-directional elastic stretch Conformable to body contours and curves Grid liner enables precise cutting and customization to any shape or size Water resistant 5 centimeters x 5 meters
Catalog No. 50-283-1437 Supplier Med Vet International Supplier No. 1019019CS
Reliable securement in damp areas; suitable for wet or diaphoretic skin Bi-directional elastic stretch Conformable to body contours and curves Grid liner enables precise cutting and customization to any shape or size Water resistant 5 centimeters x 5 meters