Learn More
Med Vet International Perforated Medical Tape 3M Medipore H White 1 inch X 2 Yard Soft Cloth Nonsterile, 2860S-1, 72/Case
Catalog No. 502832151
Encompass_Preferred
3M Medipore H surgical tape contains bi-directional stretch that adjusts to swelling and distention It won't restrict movement so it's suitable for many areas of the body 3M Medipore H medical tape is soft and breathable, providing protection that assists with wound healing It comes wrapped to lower the risk of cross-contamination It's perforated, eliminating the need for scissors or measuring There are six dozen rolls of soft cloth medical tape in a case
Catalog No. 50-283-2151 Supplier Med Vet International Supplier No. 1201191CS
3M Medipore H surgical tape contains bi-directional stretch that adjusts to swelling and distention It won't restrict movement so it's suitable for many areas of the body 3M Medipore H medical tape is soft and breathable, providing protection that assists with wound healing It comes wrapped to lower the risk of cross-contamination It's perforated, eliminating the need for scissors or measuring There are six dozen rolls of soft cloth medical tape in a case