Learn More
Med Vet International Dynarex Surgical Tape, Transparent, 3" X 10 Yards, 48/Case
Catalog No. 502829553
Encompass_Preferred
Clear surgical tape is stretchable, porous, and adhesive. Stretches and conforms to body contours Ideal for IV tubing and catheter tape downs Superior adhesion Highly porous Not made with natural rubber latex
Catalog No. 50-282-9553 Supplier Med Vet International Supplier No. 3574
Clear surgical tape is stretchable, porous, and adhesive. Stretches and conforms to body contours Ideal for IV tubing and catheter tape downs Superior adhesion Highly porous Not made with natural rubber latex