Learn More
Med Vet International 3M Transpore Medical Tape, Transparent, 2 inch X 10 Yard, Plastic, Nonsterile, 1527-2, 6/Box
Catalog No. 502829817
Encompass_Preferred
Solvent-free adhesive adheres to skin and tubing Perforations allow it to tear easily for quick application This 2 inch x 10 yard tape is easy to use while wearing gloves Water-resistance allows it to adhere in wet conditions The porous material enhances breathability This hypoallergenic tape is latex-free Each box has 6 rolls of the tape
Catalog No. 50-282-9817 Supplier Med Vet International Supplier No. 5764BX
Solvent-free adhesive adheres to skin and tubing Perforations allow it to tear easily for quick application This 2 inch x 10 yard tape is easy to use while wearing gloves Water-resistance allows it to adhere in wet conditions The porous material enhances breathability This hypoallergenic tape is latex-free Each box has 6 rolls of the tape