Learn More
Med Vet International 3M Transpore Medical Tape, Transparent, 1 inch X 10 Yard, Plastic, Nonsterile, 1527-1, 12/Box
Catalog No. 502831660
Encompass_Preferred
The 3M Transpore surgical tape features a solvent-free adhesive that adheres to skin and tubing Perforations allow it to tear easily for quick application This 1-inch x 10-yard tape is easy to use while wearing gloves because it won't stick to gloves Water-resistance allows it to adhere in wet conditions The porous material enhances breathability
Catalog No. 50-283-1660 Supplier Med Vet International Supplier No. 5762BX
The 3M Transpore surgical tape features a solvent-free adhesive that adheres to skin and tubing Perforations allow it to tear easily for quick application This 1-inch x 10-yard tape is easy to use while wearing gloves because it won't stick to gloves Water-resistance allows it to adhere in wet conditions The porous material enhances breathability