Learn More
Med Vet International Medical Tape 3M Transpore Transparent 1/2 inch X 10 Yard Plastic Nonsterile, 1527-0, 240/Case
Catalog No. 502829818
Encompass_Preferred
Solvent-free adhesive adheres to skin and tubing Perforations allow it to tear easily for quick application Transparent tape is easy to use while wearing gloves because it won't stick to gloves Water-resistance allows it to adhere in wet conditions The porous material enhances breathability 1/2 inch x 10 yards
Catalog No. 50-282-9818 Supplier Med Vet International Supplier No. 5761CS
Solvent-free adhesive adheres to skin and tubing Perforations allow it to tear easily for quick application Transparent tape is easy to use while wearing gloves because it won't stick to gloves Water-resistance allows it to adhere in wet conditions The porous material enhances breathability 1/2 inch x 10 yards