Learn More
Enterprise Technology Solutions Super 33+ Vinyl Electrical Tape, 1" Core, 0.5" X 5.5 Yds, Black
SDP
Catalog No. 502052396
Encompass_Preferred
Scotch Super 33+ Vinyl Electrical Tape is among the highest-quality electrical tapes available. Highly conformable and super stretchy in all weather applications, it resists UV rays, abrasion, corrosion, alkalis and acids. May be used as primary insulation for splices up to 600V. Suitable for indoor and outdoor use.
Catalog No. 50-205-2396 Supplier Enterprise Technology Solutions Supplier No. MMM194
Scotch Super 33+ Vinyl Electrical Tape is among the highest-quality electrical tapes available. Highly conformable and super stretchy in all weather applications, it resists UV rays, abrasion, corrosion, alkalis and acids. May be used as primary insulation for splices up to 600V. Suitable for indoor and outdoor use.