Learn More
Fisherfinest™ Taped Biopsy Cassettes for Leica IP C and Sakura Cassette Printers
Catalog No. 23021570 Shop All Fisherbrand Products
Encompass
Change view
Click to view available options
Color:
Lilac
Catalog No. Color
23-021-570 Lilac
1 options
This item is not returnable. View return policy
Catalog No. 23-021-570
This item is not returnable. View return policy
Pre-stacked, quick-load cassettes designed with efficient MicrOflow II™ reagent exchange technology
  • Validated by Leica Applications Lab for efficient, error-free use with Leica IP C and Sakura cassette printers
  • Taped, pre-stack delivery method provides quick and efficient loading of the cassette magazines
  • One-piece cassettes with frangible hinge for easy lid removal
  • 45-Degree print surface for optimal workflow
  • 0.67mm Pore
  • Available in 6 popular colors that are optimized for use with 2D barcode scanning
Patented MicrOflow II™ technology features lateral vent design for maximum fluid exchange and efficient tissue processing, as well as a unique design with angled interior for easy access to the specimen.
TRUSTED_SUSTAINABILITY
Specifications
Writing Face Angle 45°
Color Lilac
Product Type Biopsy Cassette