Learn More
Fisherfinest™ Taped Embedding Cassettes for Leica IP C and Sakura Cassette Printers
Shop All Fisherbrand Products
Description
- Validated by Leica Applications Lab for efficient, error-free use with Leica IP C and Sakura cassette printers
- Taped, pre-stack delivery method provides quick and efficient loading of the cassette magazines
- One-piece cassettes with frangible hinge for easy lid removal
- 45-Degree print surface for optimal workflow
- Available in 6 popular colors that are optimized for use with 2D barcode scanning
Patented TurbOflow II™ technology features lateral vent design for maximum fluid exchange and improved tissue processing.
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Pink |
| Product Type | Embedding Cassette |