Learn More
MSC ALPHATEC CHEM RESIST JCKT
Supplier:  MSC 41591249
Encompass
Disposable & Chemical Resistant Lab Coats; Garment Style: Jacket ; Gender: Unisex ; Size: 5X-Large ; Cuff Style: Elastic ; Material: Multi-Layer Non-Woven Barrier Laminate Fabric ; Closure Type: Double Zipper
Catalog No. 50-273-8783