Learn More
Essendant General-Purpose Masking Tape, 3"" Core, 48 mm x 54.8 m, Beige, 2/Pack
Supplier:  Essendant UNV51302
Encompass_Preferred
Beige crepe paper masking tape coated with an aggressive pressure-sensitive natural rubber adhesive. Designed primarily as an all-purpose product for non-critical applications. Ideal for interior paint masking, textile, carpet seaming, silk screening, mail rooms, golf club shaft (build-up), and for light-duty bundling, packaging, industrial, manufacturing, splicing, and tabbing. Tears easily. Lays down flat and won't lift or curl. Removes easily without leaving a residue.
Catalog No. 26-776-31