Learn More
Premier Lab Supply Tapered Wall Aluminum Cups Producing 40mm Pellets – PN# AC-540
SDP
Catalog No. NC1875128
Encompass_Preferred
Tapered Wall Aluminum Cups with Dimensions 39.8mm x 9.2mm (PN# AC-540) – Suitable for producing 40mm pellets – PREMIER’s aluminum cups are made with a strong and durable aluminum alloy that provides stable backbone for pressed pellets. Thin wall material ensures minimal back pressure and eases reproducibility of pressed pellets. – 600 per bag.
Catalog No. NC1875128 Supplier Premier Lab Supply Supplier No. AC540
Tapered Wall Aluminum Cups with Dimensions 39.8mm x 9.2mm (PN# AC-540) – Suitable for producing 40mm pellets – PREMIER’s aluminum cups are made with a strong and durable aluminum alloy that provides stable backbone for pressed pellets. Thin wall material ensures minimal back pressure and eases reproducibility of pressed pellets. – 600 per bag.