Learn More
Cole-Parmer Cole-Parmer Threaded Ultrasonic Probe Tip, Tapered Microtip; 1/8" Dia
Catalog No. NC9018808 Shop All Cole-Parmer Products
Encompass_Preferred
These microtips can process small samples at very high intensity. The tapered microtip screws in the 1/2 inch (13 mm) threaded end probe in place of the replaceable tip.
Catalog No. NC9018808 Supplier Cole-Parmer Supplier No. 0471042
These microtips can process small samples at very high intensity. The tapered microtip screws in the 1/2 inch (13 mm) threaded end probe in place of the replaceable tip.