Learn More
Glass Expansion Tapered Alumina Injector 1.5mm
SDP
Supplier:  Glass Expansion 318083202
Encompass_Preferred
Tapered Alumina Injector 1.5mm. Compatible with Glass Expansion’s Semi Demountable Torch for Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES (30-808-4335). Alumina injectors are HF resistant and are recommended for use with corrosive acids.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3142