Learn More
Glass Expansion Tapered Alumina Injector 1.8mm
SDP
Supplier:  Glass Expansion 318080814
Encompass_Preferred
Alumina Tapered Injector with 1.8mm ID.A 1.8mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and less volatile organic solvent-based matrices. Compatible with Agilent (Varian) ICP-OES Vista/700ES Series Axial Semi DemountableTorch (SDT) 30-808-0383.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7111