Learn More
Glass Expansion Tapered Alumina Injector 2.0mm
SDP
Supplier:  Glass Expansion 318070060
Encompass_Preferred
Alumina Tapered Injector with 2.0mm ID. A 2.0mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, matrices. Compatible with PerkinElmer ICP-MS Elan (non DRC), NexION 300/350 D-Torch (30-807-0504/30-808-2937) and standard torch. Suitable for torch adaptor 31-808-2825. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7055