Learn More
Glass Expansion Tapered Alumina Injector 2.4mm
SDP
Supplier:  Glass Expansion 318080401
Encompass_Preferred
Tapered Alumina Injector with 2.4mm ID. 2.4mm ID Tapered Alumina Injector. The most versatile and suitable for use with aqueous, including HF, and organic solvent-based matrices. Compatible with Agilent (Varian) 700-ES or Vista Axial Semi Demountable Torch 30-808-0379, which is obsolete.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7086