Learn More
Glass Expansion Tapered Alumina Injector 1.5mm
SDP
Catalog No. 501907061
Encompass_Preferred
Tapered Alumina Injector with 1.5mm ID. 1.5mm ID Tapered Alumina Injector. The most versatile and suitable for use with aqueous, including HF, and organic solvent-based matrices. Compatible with Spectro Blue EOP D-Torch 50-190-7034 (30-808-3600), Blue TI D-Torch 50-190-7036 (30-808-3629) and Arcos II SOP, Blue SOP & Green SOP/DSOI D-Torch 50-190-7035 (30-808-3614). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7061 Supplier Glass Expansion Supplier No. 318080013
Tapered Alumina Injector with 1.5mm ID. 1.5mm ID Tapered Alumina Injector. The most versatile and suitable for use with aqueous, including HF, and organic solvent-based matrices. Compatible with Spectro Blue EOP D-Torch 50-190-7034 (30-808-3600), Blue TI D-Torch 50-190-7036 (30-808-3629) and Arcos II SOP, Blue SOP & Green SOP/DSOI D-Torch 50-190-7035 (30-808-3614). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001