Learn More
Glass Expansion Tapered Alumina Injector 2.4mm for D-Torch
SDP
Catalog No. 501907188
Encompass_Preferred
Alumina Tapered Injector with 2.4mm ID.A 2.4mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with PerkinElmer ICP-OES NexION 1000/2000/5000 (30-808-3927). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7188 Supplier Glass Expansion Supplier No. 318083205
Alumina Tapered Injector with 2.4mm ID.A 2.4mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with PerkinElmer ICP-OES NexION 1000/2000/5000 (30-808-3927). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001