Learn More
Glass Expansion Tapered Alumina Injector 2.4mm
SDP
Supplier:  Glass Expansion 318083855
Encompass_Preferred
Alumina Tapered Injector with 2.4mm ID.A 2.4mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with Agilent ICP-MS 7700/7800/7900/8800/8900 Semi Demountable Torch Body NC2046979 (30-808-3857).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7264