Learn More
Glass Expansion Tapered Alumina Injector 2.4mm
SDP
Supplier:  Glass Expansion 318083700
Encompass_Preferred
Tapered Alumina Injector with 2.4mm ID.2.4mm ID Tapered Alumina Injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with Spectro Arcos II EOP D-Torch (30-808-3657).When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7248