Learn More
Glass Expansion Tapered Alumina Injector 2.4mm
SDP
Catalog No. 501907251
Encompass_Preferred
Alumina Tapered Injector with 2.4mm ID.A 2.4mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with Agilent ICP-OES 5100/5110/5800/5900 D-Torch (30-808-3560). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7251 Supplier Glass Expansion Supplier No. 318083722
Alumina Tapered Injector with 2.4mm ID.A 2.4mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with Agilent ICP-OES 5100/5110/5800/5900 D-Torch (30-808-3560). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001