Learn More
Glass Expansion Tapered Alumina Injector 1.8mm
SDP
Catalog No. 501907235
Encompass_Preferred
Alumina Tapered Injector with 1.8mm ID.A 1.8mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and less volatile organic solvent-based matrices. Compatible with Agilent ICP-OES 5100/5110/5800/5900 D-Torch (30-808-3560). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7235 Supplier Glass Expansion Supplier No. 318083585
Alumina Tapered Injector with 1.8mm ID.A 1.8mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and less volatile organic solvent-based matrices. Compatible with Agilent ICP-OES 5100/5110/5800/5900 D-Torch (30-808-3560). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001