Learn More
Glass Expansion Tapered Alumina Injector 2.0mm
SDP
Catalog No. 501907247
Encompass_Preferred
Tapered Alumina Injector with 2.0mm ID.2.0mm ID Tapered Alumina Injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with Spectro Arcos II EOP D-Torch (30-808-3657).When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7247 Supplier Glass Expansion Supplier No. 318083699
Tapered Alumina Injector with 2.0mm ID.2.0mm ID Tapered Alumina Injector. The most versatile and suitable for use with aqueous, including HF, and high TDS matrices. Compatible with Spectro Arcos II EOP D-Torch (30-808-3657).When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001