Learn More
Glass Expansion Tapered Alumina Injector 1.5mm
SDP
Supplier:  Glass Expansion 318083725
Encompass_Preferred
Alumina Tapered Injector with 1.5mm ID.A 1.5mm ID alumina tapered injector. The most versatile and suitable for use with aqueous, including HF, and organic solvent-based matrices. Compatible with Agilent ICP-OES 5100/5110/5800/5900 Series D-Torch 50-190-7032 (30-808-3590) and 30-808-3560. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7253