Learn More
Ace Glass, Inc. Tapered flask, 50mL, 3 neck, 14/20 center, (2) 14/20
SDP
Catalog No. 50976904
Encompass_Preferred
50mL three neck tapered flask with walls tapered inward toward the bottom to allow continuous operation with smaller volumes. Flask has 14/20 center neck and (2) 14/20 side necks.
Catalog No. 50-976-904 Supplier Ace Glass, Inc. Supplier No. 947330
50mL three neck tapered flask with walls tapered inward toward the bottom to allow continuous operation with smaller volumes. Flask has 14/20 center neck and (2) 14/20 side necks.