Learn More
Ace Glass, Inc. Tapered flask, 125mL, 4 neck, 14/20 center, (2) 14/20, (1) #7Ace-Thred
SDP
Catalog No. 50976903
Encompass_Preferred
125mL four neck tapered flask with walls tapered inward toward the bottom to allow continuous operation with smaller volumes. Flask has 14/20 center neck, (2) 14/20 side necks, and (2) #7 Ace-Thred necks. #7 Ace-Thred necks can accomm ODate thermometers and electrodes up to 7mm O.D. Ace-Threds include nylon bushings and FETFE o-rings.
Catalog No. 50-976-903 Supplier Ace Glass, Inc. Supplier No. 947324
125mL four neck tapered flask with walls tapered inward toward the bottom to allow continuous operation with smaller volumes. Flask has 14/20 center neck, (2) 14/20 side necks, and (2) #7 Ace-Thred necks. #7 Ace-Thred necks can accomm ODate thermometers and electrodes up to 7mm O.D. Ace-Threds include nylon bushings and FETFE o-rings.