Learn More
Sigma Aldrich Fine Chemicals Biosciences Tapered microtip for 500 a1EA
Catalog No. 502905073
Encompass
Screw into threaded end of standard 1/2 in. (13 mm) horn to intensify the energy delivered to small samples. Also for use with narrow neck vessels.
Catalog No. 50-290-5073 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1927401EA
Screw into threaded end of standard 1/2 in. (13 mm) horn to intensify the energy delivered to small samples. Also for use with narrow neck vessels.