Learn More
Glass Expansion Tapered Quartz Injector 1.0mm
SDP
Catalog No. 501907068
Encompass_Preferred
Quartz Tapered Injector with 1.0mm ID. 1.0mm ID tapered quartz injector. This quartz injector is for use with PerkinElmer Optima 4300V/5300V/7300V Series ICP-OES instruments. They are designed for organic solvents and non-HF matrices. A variety of sizes are available as an alternative to the standard injector.PerkinElmer Optima 4300V/5300V/7300V ICP-OES Series Injectors - Compatible with 50-190-7156 (31-808-2821), 50-190-6981 (30-807-0064), 50-190-7033 (30-808-3593), 50-190-7020 (30-808-2832), 50-210-4733 (N0771561), 50-904-3809 (N0771500). Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7068 Supplier Glass Expansion Supplier No. 318080119
Quartz Tapered Injector with 1.0mm ID. 1.0mm ID tapered quartz injector. This quartz injector is for use with PerkinElmer Optima 4300V/5300V/7300V Series ICP-OES instruments. They are designed for organic solvents and non-HF matrices. A variety of sizes are available as an alternative to the standard injector.PerkinElmer Optima 4300V/5300V/7300V ICP-OES Series Injectors - Compatible with 50-190-7156 (31-808-2821), 50-190-6981 (30-807-0064), 50-190-7033 (30-808-3593), 50-190-7020 (30-808-2832), 50-210-4733 (N0771561), 50-904-3809 (N0771500). Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001