Learn More
Glass Expansion Tapered Quartz Injector 2.0mm
SDP
Supplier:  Glass Expansion 318070004
Encompass_Preferred
Quartz Tapered Injector with 2.0mm ID. A 2.0mm ID quartz tapered injector suitable for use with aqueous matrices, including samples with high TDS. Compatible with PerkinElmer ICP-MS Elan (non DRC), NexION 300/350 D-Torch (30-808-3927) and standard torch. Not for use with HF. Suitable for torch adaptor 31-808-2825. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7046