Learn More
Glass Expansion Tapered Quartz Injector 1.0mm
SDP
Supplier:  Glass Expansion 318080296
Encompass_Preferred
Quartz Tapered Injector with 1.0mm ID. A 1.0mm ID quartz tapered injector, suitable for use with volatile organic solvent-based matrices. Compatible with PT2000 TJA Axial Torch, which is obsolete. Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7076