Learn More
Glass Expansion Tapered Quartz Injector 1.5mm
SDP
Supplier:  Glass Expansion 318080924
Encompass_Preferred
Quartz Tapered Injector with 1.5mm ID. A 1.5mm ID quartz tapered injector, ideal for use with volatile organic solvent-based and non-HF matrices. Compatible with Nu Plasma II ICP-MS D-Torch 30-808-3496. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7122