Learn More
Glass Expansion Tapered Quartz Injector 1.5mm
SDP
Supplier:  Glass Expansion 318070006
Encompass_Preferred
Quartz Tapered Injector with 1.5mm ID. A 1.5mm ID quartz tapered injector, ideal for use with volatile organic solvent-based and non-HF aqueous matrices. Compatible with PerkinElmer ICP-MS Elan (non DRC), NexION 300/350 D-Torch (30-808-3927) and standard torch. Not for use with HF. Suitable for torch adaptor 31-808-2825. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7048