Learn More
Glass Expansion Tapered Quartz Injector 1.0mm
SDP
Supplier:  Glass Expansion 318082719
Encompass_Preferred
Quartz Tapered Injector with 1.0mm ID for old design torch (842312051341).A 1.0mm ID quartz tapered injector for old design torch. Ideal for use with volatile organic solvent-based matrices. Not for use with HF.Compatible with Thermo 6000/7000 Duo old design torch (31-808-2676), and Thermo 6000/7000 Radial old design torch (842312051141 and 31-808-2692).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7140