Learn More
Glass Expansion Tapered Quartz Injector for D-Torch 2.4mm
SDP
Catalog No. 501907174
Encompass_Preferred
Quartz Tapered Injector with 2.4mm ID. A 2.4mm ID quartz tapered injector suitable for use with aqueous matrices. Compatible with Agilent ICP-OES 700-ES Series Axial, Vista Axial D-Torch (501907021).Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7174 Supplier Glass Expansion Supplier No. 318083043
Quartz Tapered Injector with 2.4mm ID. A 2.4mm ID quartz tapered injector suitable for use with aqueous matrices. Compatible with Agilent ICP-OES 700-ES Series Axial, Vista Axial D-Torch (501907021).Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001