Learn More
Glass Expansion Tapered Quartz Injector for D-Torch 1.5mm
SDP
Catalog No. 501907176
Encompass_Preferred
Quartz Tapered Injector with 1.5mm ID. A 1.5mm ID quartz tapered injector suitable for use with aqueous and organic solvent-based matrices. Compatible with Agilent ICP-OES 700-ES Series Radial, Vista Radial D-Torch (501907026). Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7176 Supplier Glass Expansion Supplier No. 318083058
Quartz Tapered Injector with 1.5mm ID. A 1.5mm ID quartz tapered injector suitable for use with aqueous and organic solvent-based matrices. Compatible with Agilent ICP-OES 700-ES Series Radial, Vista Radial D-Torch (501907026). Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001