Learn More
Glass Expansion Tapered Quartz Injector 2.5mm
SDP
Catalog No. 501907246
Encompass_Preferred
Tapered Quartz Injector with 2.5mm ID.2.5mm ID Tapered Quartz Injector. They are designed for organic solvents and non-HF matrices. Compatible with Spectro Arcos II EOP D-Torch (30-808-3657).When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7246 Supplier Glass Expansion Supplier No. 318083676
Tapered Quartz Injector with 2.5mm ID.2.5mm ID Tapered Quartz Injector. They are designed for organic solvents and non-HF matrices. Compatible with Spectro Arcos II EOP D-Torch (30-808-3657).When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001