Learn More
Glass Expansion Tapered Quartz Injector 2.5mm for D-Torch
SDP
Supplier:  Glass Expansion 318083723
Encompass_Preferred
Tapered Quartz Injector with 2.5mm ID.2.5mm ID Tapered Quartz Injector. They are designed for organic solvents and non-HF matrices. Compatible with Spectro (Ametek) SOP Modula, CIROS, Genesis, ARCOS Radial D-Torch 50-190-7023 (30-808-2928). When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7252