Learn More
Glass Expansion Tapered Quartz Injector for D-Torch 1.5mm with Ball Joint
SDP
Catalog No. 501907184
Encompass_Preferred
Quartz Tapered Injector 1.5mm ID with Ball Joint. A 1.5mm ID quartz tapered injector with ball joint suitable for use with aqueous and organic solvent-based matrices. Compatible with Agilent ICP-OES 700-ES Series Radial, Vista Radial D-Torch (501907026). Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7184 Supplier Glass Expansion Supplier No. 318083150
Quartz Tapered Injector 1.5mm ID with Ball Joint. A 1.5mm ID quartz tapered injector with ball joint suitable for use with aqueous and organic solvent-based matrices. Compatible with Agilent ICP-OES 700-ES Series Radial, Vista Radial D-Torch (501907026). Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001