Learn More
Glass Expansion Tapered Quartz Injector with ball joint 2.5mm (suitable for Elan DRC/NexION)
SDP
Catalog No. 501907224
Encompass_Preferred
Quartz Tapered Injector with Ball Joint 2.5mm ID. A 2.5mm ID quartz tapered injector with ball joint for use with aqueous matrices, including samples with high TDS. Compatible with PerkinElmer ICP-MS Elan DRC, NexION 300/350 D-Torch (501906989) and standard torch. Not for use with HF. Suitable for torch adaptor 31-808-2824. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7224 Supplier Glass Expansion Supplier No. 318083463
Quartz Tapered Injector with Ball Joint 2.5mm ID. A 2.5mm ID quartz tapered injector with ball joint for use with aqueous matrices, including samples with high TDS. Compatible with PerkinElmer ICP-MS Elan DRC, NexION 300/350 D-Torch (501906989) and standard torch. Not for use with HF. Suitable for torch adaptor 31-808-2824. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001