Learn More
Glass Expansion Tapered Quartz Injector 2.0mm
SDP
Catalog No. 501907196
Encompass_Preferred
Quartz Tapered Injector with 2.0mm ID. 2.0mm ID tapered quartz injector (N0781050). This quartz Injector is for use with Optima 8x00 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices. Optima 8x00 Series Injectors - Compatible with 50-190-7027 (30-808-3300), 50-190-7193 (31-808-3254), 50-190-7194 (31-808-3255), 50-190-7195 (31-808-3256), 50-211-2265 (N0780132), 50-211-2268 (N0780133), 50-211-2814 (N0780119).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7196 Supplier Glass Expansion Supplier No. 318083258
Quartz Tapered Injector with 2.0mm ID. 2.0mm ID tapered quartz injector (N0781050). This quartz Injector is for use with Optima 8x00 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices. Optima 8x00 Series Injectors - Compatible with 50-190-7027 (30-808-3300), 50-190-7193 (31-808-3254), 50-190-7194 (31-808-3255), 50-190-7195 (31-808-3256), 50-211-2265 (N0780132), 50-211-2268 (N0780133), 50-211-2814 (N0780119).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001