Learn More
Glass Expansion Tapered Quartz Injector 2.5mm for D-Torch
SDP
Catalog No. 501907205
Encompass_Preferred
Quartz Tapered Injector with 2.5mm ID. A 2.5mm ID quartz tapered injector suitable for use with aqueous matrices, including samples with high TDS. Compatible with PerkinElmer ICP-MS NexION 1000/2000/5000 D-Torch (30-808-3927). Not suitable for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7205 Supplier Glass Expansion Supplier No. 318083302
Quartz Tapered Injector with 2.5mm ID. A 2.5mm ID quartz tapered injector suitable for use with aqueous matrices, including samples with high TDS. Compatible with PerkinElmer ICP-MS NexION 1000/2000/5000 D-Torch (30-808-3927). Not suitable for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001