Learn More
Glass Expansion Tapered Quartz Injector 1.8mm
SDP
Supplier:  Glass Expansion 318088917
Encompass_Preferred
Quartz Tapered Injector with 1.8mm ID. A 1.8mm ID quartz tapered injector (14110500), suitable for use with aqueous and less volatile organic solvent-based matrices. Compatible with Horiba Jobin Yvon ICP-OES 24, 38 & 138 Series, Ultima, Activa PT2000 Fully Demountable Torch, which is obsolete. Not suitable for use with HF.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7278