Learn More
Glass Expansion Tapered Quartz Injector 1.8mm
SDP
Supplier:  Glass Expansion 318088146
Encompass_Preferred
Tapered Quartz Injector with 1.8mm ID.1.8mm ID Tapered Quartz Injector. They are designed for organic solvents and non-HF matrices. Compatible with Spectro Blue EOP D-Torch 50-190-7034 (30-808-3600), Blue TI D-Torch 50-190-7036 (30-808-3629) and Arcos II SOP, Blue SOP & Green SOP/DSOI D-Torch 50-190-7035 (30-808-3614).When replacing your injector, it is recommended to replace the injector ferrule (31-808-2962), on average every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7274