Learn More
Glass Expansion Tapered Quartz Injector 1.8mm
SDP
Catalog No. 501907047
Encompass_Preferred
Quartz Tapered Injector with 1.8mm ID. A 1.8mm ID quartz tapered injector suitable for use with aqueous and less volatile organic solvent-based matrices. Compatible with PerkinElmer ICP-MS Elan (non DRC), NexION 300/350 D-Torch (30-808-3927) and standard torch. Not for use with HF. Suitable for torch adaptor 31-808-2825. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7047 Supplier Glass Expansion Supplier No. 318070005
Quartz Tapered Injector with 1.8mm ID. A 1.8mm ID quartz tapered injector suitable for use with aqueous and less volatile organic solvent-based matrices. Compatible with PerkinElmer ICP-MS Elan (non DRC), NexION 300/350 D-Torch (30-808-3927) and standard torch. Not for use with HF. Suitable for torch adaptor 31-808-2825. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001