Learn More
Electron Microscopy Sciences Tapered Tissue Grinder, 1 ml, 11 mm x 49 mm Mortor, 130 mm OAL
SDP
Catalog No. 501900348
Encompass_Preferred
This is new design grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require. The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step. It is ideal for homogenization of connective tissue including heart, muscle and lung, as well as skin and plant tissue.
Made from Wheaton 33 low extractable borosilicate glass. Interchangeable components takes the hassle out of matching parts. Unit is autoclavable. When the pestle is "bottomed" within the mortar, the clearance is uniform 0.004" to 0.006” (0.1 to 0.15 mm).
Size: 1 ml
Mortor: 11 mm x 49 mm
OAL: 130 mm
Catalog No. 50-190-0348 Supplier Electron Microscopy Sciences Supplier No. 6479601
This is new design grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require. The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step. It is ideal for homogenization of connective tissue including heart, muscle and lung, as well as skin and plant tissue.
Made from Wheaton 33 low extractable borosilicate glass. Interchangeable components takes the hassle out of matching parts. Unit is autoclavable. When the pestle is "bottomed" within the mortar, the clearance is uniform 0.004" to 0.006” (0.1 to 0.15 mm).
Size: 1 ml
Mortor: 11 mm x 49 mm
OAL: 130 mm