Learn More
Electron Microscopy Sciences Tapered Tissue Grinder with Stainless Steel Rod PTFE Pestle, 1 ml, Mortor, 130 mm OAL
SDP
Catalog No. 50365139
Encompass_Preferred
This is new design grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require. The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step. Made from Wheaton 33 low extractable borosilicate glass. Interchangeable components takes the hassle out of matching parts. Unit is autoclavable. When the pestle is "bottomed" within the mortar, the clearance is uniform 0.004" to 0.006” (0.1 to 0.15 mm). Teflon pestle with stainless steal rod firs power homogenizer. Homogenizes soft tissue such as brain. The stainless steel rod is 0.25” (6.3 mm) diameter and is suited for use with Wheaton Overhead Stirrer.
Size: 1 ml
OAL: 130 mm
Catalog No. 50-365-139 Supplier Electron Microscopy Sciences Supplier No. 6479701
This is new design grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require. The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step. Made from Wheaton 33 low extractable borosilicate glass. Interchangeable components takes the hassle out of matching parts. Unit is autoclavable. When the pestle is "bottomed" within the mortar, the clearance is uniform 0.004" to 0.006” (0.1 to 0.15 mm). Teflon pestle with stainless steal rod firs power homogenizer. Homogenizes soft tissue such as brain. The stainless steel rod is 0.25” (6.3 mm) diameter and is suited for use with Wheaton Overhead Stirrer.
Size: 1 ml
OAL: 130 mm