Learn More
Electron Microscopy Sciences Tapered Tissue Grinder, 3 ml, 11 mmx 86 mm Mortor, 220 mm OAL
SDP
Catalog No. 501900349
Encompass_Preferred
This is new design grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require. The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step. It is ideal for homogenization of connective tissue including heart, muscle and lung, as well as skin and plant tissue.
Made from Wheaton 33 low extractable borosilicate glass. Interchangeable components takes the hassle out of matching parts. Unit is autoclavable. When the pestle is "bottomed" within the mortar, the clearance is uniform 0.004" to 0.006” (0.1 to 0.15 mm).
Size: 3 ml
Mortor: 11 mmx 86 mm
OAL: 220 mm
Catalog No. 50-190-0349 Supplier Electron Microscopy Sciences Supplier No. 6479603
This is new design grinder with longer tapered surface on both the mortar and pestle, offering grinding more efficient and less time require. The conical surface allows initial size reduction followed by passage through the cylindrical section for the final homogenization step. It is ideal for homogenization of connective tissue including heart, muscle and lung, as well as skin and plant tissue.
Made from Wheaton 33 low extractable borosilicate glass. Interchangeable components takes the hassle out of matching parts. Unit is autoclavable. When the pestle is "bottomed" within the mortar, the clearance is uniform 0.004" to 0.006” (0.1 to 0.15 mm).
Size: 3 ml
Mortor: 11 mmx 86 mm
OAL: 220 mm