Learn More
Electron Microscopy Sciences Tapered Tissue Mortar only, 15 ml, 18 mm x 114 mm Mortor, 240 mm OAL
SDP
Catalog No. 501901624
Encompass_Preferred
Replacement parts for Tapered Tissue Grinder with stainless steel rod pestle.
Size: 15 ml
Mortor: 18 mm x 114 mm
OAL: 240 mm
Catalog No. 50-190-1624 Supplier Electron Microscopy Sciences Supplier No. 6479773
Replacement parts for Tapered Tissue Grinder with stainless steel rod pestle.
Size: 15 ml
Mortor: 18 mm x 114 mm
OAL: 240 mm