Learn More
Electron Microscopy Sciences Tapered Tissue Mortar only, 3 ml, 11 mm x 86 mm Mortor, 220 mm OAL
SDP
Catalog No. 501901622
Encompass_Preferred
Replacement parts for Tapered Tissue Grinder with stainless steel rod pestle.
Size: 3 ml
Mortor: 11 mm x 86 mm
OAL: 220 mm
Catalog No. 50-190-1622 Supplier Electron Microscopy Sciences Supplier No. 6479771
Replacement parts for Tapered Tissue Grinder with stainless steel rod pestle.
Size: 3 ml
Mortor: 11 mm x 86 mm
OAL: 220 mm